Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THYN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204287.2 | NP_989618.1 | 4 - 5 | 485 | 76 | |
XM_015297937.2 | XP_015153423.1 | 4 - 5 | 496 | 76 | ||
GenBank mRNA | BX950750.2 | - | 4 - 5 | 484 | 76 | |
U34350.1 | - | 4 - 5 | 485 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |