Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204305.1 | NP_989636.1 | 10 - 11 | 888 | 76 | |
GenBank mRNA | AF047874.1 | - | 10 - 11 | 888 | 76 | |
AY466166.1 | - | 3 - 4 | 247 | 76 | ||
J00850.1 | - | 8 - 9 | 765 | 76 | ||
K01458.1 | - | 10 - 11 | 887 | 76 | ||
V00407.1 | - | 9 - 10 | 860 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Inventoried Probe spans exons |