Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204376.2 | NP_989707.1 | 4 - 5 | 391 | 98 | |
GenBank mRNA | AJ720701.1 | - | 4 - 5 | 391 | 98 | |
AY336970.1 | - | 4 - 5 | 391 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |