Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204388.2 | NP_989719.2 | 4 - 5 | 1138 | 100 | |
XM_015285869.1 | XP_015141355.1 | 4 - 5 | 1205 | 100 | ||
XM_015285870.2 | XP_015141356.1 | 4 - 5 | 1131 | 100 | ||
GenBank mRNA | AF396666.1 | - | 3 - 4 | 925 | 100 | |
AJ720049.1 | - | 4 - 5 | 1138 | 100 | ||
AJ851790.1 | - | 4 - 5 | 1128 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |