Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204541.2 | NP_989872.1 | 4 - 5 | 486 | 99 | |
GenBank mRNA | AB085851.1 | - | 4 - 5 | 445 | 99 | |
AY584568.1 | - | 4 - 5 | 486 | 99 | ||
BX934742.2 | - | 4 - 5 | 495 | 99 | ||
EU049890.1 | - | 4 - 5 | 443 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |