Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204817.1 | NP_990148.1 | 2 - 3 | 439 | 68 | |
XM_015284813.2 | XP_015140299.1 | 2 - 3 | 590 | 68 | ||
GenBank mRNA | AJ131923.1 | - | 2 - 3 | 439 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |