Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204910.1 | NP_990241.1 | 4 - 5 | 477 | 73 | |
GenBank mRNA | AF065629.1 | - | 4 - 5 | 477 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |