Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204932.1 | NP_990263.1 | 2 - 3 | 657 | 68 | |
GenBank mRNA | AB734688.1 | - | 2 - 3 | 600 | 68 | |
AB734689.1 | - | 2 - 3 | 600 | 68 | ||
AF072698.1 | - | 2 - 3 | 657 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |