Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204956.1 | NP_990287.1 | 9 - 10 | 1117 | 128 | |
XM_015286577.2 | XP_015142063.2 | 9 - 10 | 1189 | 128 | ||
GenBank mRNA | U26428.1 | - | 9 - 10 | 1117 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |