Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204962.1 | NP_990293.1 | 3 - 4 | 694 | 92 | |
GenBank mRNA | U44473.1 | - | 3 - 4 | 694 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |