Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP24A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204979.1 | NP_990310.1 | 9 - 10 | 1223 | 90 | |
GenBank mRNA | AF019142.1 | - | 9 - 10 | 1223 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |