Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204991.1 | NP_990322.1 | 7 - 8 | 1154 | 68 | |
XM_015286039.2 | XP_015141525.1 | 7 - 8 | 1644 | 68 | ||
XM_015286040.2 | XP_015141526.1 | 7 - 8 | 1181 | 68 | ||
GenBank mRNA | AF045162.1 | - | 7 - 8 | 1154 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |