Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205005.1 | NP_990336.1 | 3 - 4 | 234 | 94 | |
GenBank mRNA | AF081672.1 | - | 3 - 4 | 234 | 94 | |
AJ720728.1 | - | 3 - 4 | 266 | 94 | ||
M36167.1 | - | 2 - 3 | 175 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |