Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205016.1 | NP_990347.1 | 5 - 6 | 470 | 109 | |
GenBank mRNA | U79756.1 | - | 5 - 6 | 470 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |