Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205041.1 | NP_990372.1 | 1 - 2 | 189 | 108 | |
XM_015293006.2 | XP_015148492.1 | 1 - 2 | 206 | 108 | ||
GenBank mRNA | AB002585.1 | - | 1 - 2 | 189 | 108 | |
AB002586.1 | - | 1 - 2 | 189 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |