Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205051.1 | NP_990382.1 | 11 - 12 | 1967 | 143 | |
XM_015276138.2 | XP_015131624.1 | 11 - 12 | 2021 | 143 | ||
GenBank mRNA | AB004649.1 | - | 11 - 12 | 1967 | 143 | |
AB004650.1 | - | 12 - 13 | 1907 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |