Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRPAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205062.1 | NP_990393.1 | 6 - 7 | 800 | 148 | |
GenBank mRNA | AJ001912.1 | - | 6 - 7 | 800 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |