Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205162.1 | NP_990493.1 | 3 - 4 | 228 | 95 | |
GenBank mRNA | U10577.1 | - | 3 - 4 | 228 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |