Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205310.1 | NP_990641.1 | 4 - 5 | 692 | 97 | |
XM_015287304.2 | XP_015142790.1 | 5 - 6 | 719 | 97 | ||
XM_025150611.1 | XP_025006379.1 | 6 - 7 | 930 | 97 | ||
GenBank mRNA | BX932948.1 | - | 5 - 6 | 703 | 97 | |
X03509.1 | - | 692 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |