Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205320.1 | NP_990651.1 | 3 - 4 | 451 | 77 | |
GenBank mRNA | X05343.1 | - | 451 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |