Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POPDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206865.1 | NP_996747.1 | 3 - 4 | 1453 | 140 | |
GenBank mRNA | AY388621.1 | - | 3 - 4 | 1453 | 140 | |
AY388623.1 | - | 4 - 5 | 1265 | 140 | ||
CR354299.1 | - | 1 - 2 | 382 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |