Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIPOQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206991.1 | NP_996874.1 | 1 - 2 | 499 | 66 | |
XM_015276846.2 | XP_015132332.1 | 2 - 3 | 620 | 66 | ||
GenBank mRNA | AY523637.1 | - | 1 - 2 | 499 | 66 | |
KP729052.1 | - | 1 - 2 | 224 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |