Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030578.1 | NP_001025749.1 | 2 - 3 | 308 | 106 | |
GenBank mRNA | AJ851825.1 | - | 2 - 3 | 308 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |