Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030578.1 | NP_001025749.1 | 7 - 8 | 866 | 72 | |
GenBank mRNA | AJ851825.1 | - | 7 - 8 | 866 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |