Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030593.2 | NP_001025764.2 | 1 - 2 | 452 | 104 | |
GenBank mRNA | AJ720682.1 | - | 1 - 2 | 453 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |