Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030597.2 | NP_001025768.1 | 13 - 14 | 1341 | 87 | |
GenBank mRNA | AJ851498.1 | - | 13 - 14 | 1342 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |