Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030597.2 | NP_001025768.1 | 23 - 24 | 2608 | 153 | |
GenBank mRNA | AJ851498.1 | - | 23 - 24 | 2609 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |