Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030616.1 | NP_001025787.1 | 1 - 2 | 176 | 94 | |
XM_015293767.1 | XP_015149253.1 | 1 - 2 | 131 | 94 | ||
GenBank mRNA | AJ720720.1 | - | 1 - 2 | 176 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |