Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030800.1 | NP_001025971.1 | 16 - 17 | 1985 | 69 | |
XM_015301535.2 | XP_015157021.1 | 16 - 17 | 1984 | 69 | ||
GenBank mRNA | AJ851409.1 | - | 16 - 17 | 1985 | 69 | |
MH119574.1 | - | 16 - 17 | 1872 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |