Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030812.1 | NP_001025983.1 | 5 - 6 | 670 | 91 | |
GenBank mRNA | AJ719844.1 | - | 5 - 6 | 670 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |