Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030814.1 | NP_001025985.1 | 3 - 4 | 494 | 79 | |
GenBank mRNA | AJ720236.1 | - | 3 - 4 | 494 | 79 | |
CR353091.1 | - | 2 - 3 | 282 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |