Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030828.1 | NP_001025999.1 | 4 - 5 | 1241 | 88 | |
XM_015277964.2 | XP_015133450.1 | 4 - 5 | 1400 | 88 | ||
GenBank mRNA | AJ851382.1 | - | 4 - 5 | 1241 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |