Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP4B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030917.1 | NP_001026088.1 | 2345 | 62 | ||
XM_025143694.1 | XP_024999462.1 | 1634 | 62 | |||
XM_025143695.1 | XP_024999463.1 | 1626 | 62 | |||
XM_025143696.1 | XP_024999464.1 | 752 | 62 | |||
GenBank mRNA | AJ720521.1 | - | 2345 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus |