Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030923.1 | NP_001026094.1 | 542 | 75 | ||
XM_015298954.2 | XP_015154440.1 | 505 | 75 | |||
XM_015298955.2 | XP_015154441.1 | 901 | 75 | |||
XM_025143708.1 | XP_024999476.1 | 692 | 75 | |||
GenBank mRNA | AJ719313.1 | - | 542 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus |