Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030942.1 | NP_001026113.1 | 7 - 8 | 1095 | 118 | |
GenBank mRNA | AJ720953.1 | - | 7 - 8 | 1095 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |