Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031024.1 | NP_001026195.1 | 5 - 6 | 521 | 91 | |
XM_015282672.2 | XP_015138158.1 | 5 - 6 | 552 | 91 | ||
XM_025147307.1 | XP_025003075.1 | 5 - 6 | 431 | 91 | ||
XM_025147308.1 | XP_025003076.1 | 5 - 6 | 471 | 91 | ||
XM_025147309.1 | XP_025003077.1 | 5 - 6 | 481 | 91 | ||
GenBank mRNA | AJ721106.1 | - | 5 - 6 | 521 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |