Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031030.1 | NP_001026201.1 | 1174 | 98 | ||
XM_015298713.2 | XP_015154199.1 | 2758 | 98 | |||
GenBank mRNA | AJ720443.1 | - | 1174 | 98 | ||
CR388606.1 | - | 183 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus |