Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031039.1 | NP_001026210.1 | 4 - 5 | 647 | 129 | |
XM_015278082.2 | XP_015133568.1 | 5 - 6 | 859 | 129 | ||
XM_015278083.2 | XP_015133569.1 | 5 - 6 | 715 | 129 | ||
XM_015278084.2 | XP_015133570.1 | 5 - 6 | 825 | 129 | ||
XM_015278085.2 | XP_015133571.1 | 4 - 5 | 649 | 129 | ||
XM_025148656.1 | XP_025004424.1 | 6 - 7 | 905 | 129 | ||
XM_025148657.1 | XP_025004425.1 | 4 - 5 | 676 | 129 | ||
GenBank mRNA | AJ851530.1 | - | 4 - 5 | 647 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |