Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031043.1 | NP_001026214.1 | 1 - 2 | 300 | 103 | |
XM_025148668.1 | XP_025004436.1 | 2 - 3 | 4819 | 103 | ||
GenBank mRNA | AJ720620.1 | - | 1 - 2 | 300 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |