Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031054.1 | NP_001026225.1 | 4 - 5 | 264 | 140 | |
GenBank mRNA | AJ720168.1 | - | 4 - 5 | 264 | 140 | |
CR387528.1 | - | 4 - 5 | 260 | 140 | ||
CR391323.1 | - | 4 - 5 | 261 | 140 | ||
CR391716.1 | - | 4 - 5 | 252 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |