Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031093.1 | NP_001026264.1 | 3 - 4 | 381 | 74 | |
XM_015284972.2 | XP_015140458.1 | 4 - 5 | 556 | 74 | ||
XM_015284973.2 | XP_015140459.1 | 3 - 4 | 377 | 74 | ||
GenBank mRNA | AJ851423.1 | - | 3 - 4 | 381 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |