Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031094.1 | NP_001026265.1 | 5 - 6 | 507 | 77 | |
XM_015284974.1 | XP_015140460.1 | 5 - 6 | 630 | 77 | ||
XM_015284975.2 | XP_015140461.1 | 6 - 7 | 707 | 77 | ||
XM_015284977.2 | XP_015140463.1 | 5 - 6 | 631 | 77 | ||
GenBank mRNA | AJ720070.1 | - | 5 - 6 | 507 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |