Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031109.3 | NP_001026280.2 | 3 - 4 | 374 | 84 | |
XM_015295405.2 | XP_015150891.1 | 3 - 4 | 355 | 84 | ||
XM_015295406.2 | XP_015150892.1 | 3 - 4 | 498 | 84 | ||
XM_015295407.2 | XP_015150893.1 | 3 - 4 | 358 | 84 | ||
XM_025141616.1 | XP_024997384.1 | 3 - 4 | 498 | 84 | ||
GenBank mRNA | AJ719294.1 | - | 3 - 4 | 365 | 84 | |
BX934833.1 | - | 3 - 4 | 425 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |