Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031136.1 | NP_001026307.1 | 7 - 8 | 846 | 122 | |
XM_015276225.2 | XP_015131711.1 | 7 - 8 | 868 | 122 | ||
XM_015276226.2 | XP_015131712.1 | 7 - 8 | 865 | 122 | ||
GenBank mRNA | AJ720831.1 | - | 7 - 8 | 846 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |