Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031172.1 | NP_001026343.1 | 1 - 2 | 247 | 82 | |
XM_015286198.2 | XP_015141684.1 | 1 - 2 | 354 | 82 | ||
GenBank mRNA | AJ720609.1 | - | 1 - 2 | 247 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |