Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031240.1 | NP_001026411.1 | 9 - 10 | 1152 | 78 | |
GenBank mRNA | AJ720214.1 | - | 9 - 10 | 1152 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |