Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031435.1 | NP_001026606.1 | 9 - 10 | 985 | 67 | |
GenBank mRNA | AJ720800.1 | - | 9 - 10 | 985 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |