Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031475.1 | NP_001026646.1 | 5 - 6 | 689 | 108 | |
XM_015279526.2 | XP_015135012.1 | 5 - 6 | 713 | 108 | ||
GenBank mRNA | AJ719979.1 | - | 5 - 6 | 689 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |