Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031507.1 | NP_001026678.1 | 3 - 4 | 361 | 121 | |
XM_015283110.2 | XP_015138596.1 | 5 - 6 | 969 | 121 | ||
XM_015283111.2 | XP_015138597.1 | 4 - 5 | 898 | 121 | ||
GenBank mRNA | AJ719948.1 | - | 3 - 4 | 361 | 121 | |
CR391308.1 | - | 3 - 4 | 188 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |