Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NONO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031532.1 | NP_001026703.1 | 2 - 3 | 200 | 93 | |
XM_015278310.2 | XP_015133796.1 | 2 - 3 | 322 | 93 | ||
XM_015278312.2 | XP_015133798.1 | 1 - 2 | 186 | 93 | ||
XM_015278313.2 | XP_015133799.1 | 2 - 3 | 206 | 93 | ||
GenBank mRNA | AJ720639.1 | - | 2 - 3 | 200 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |